KEY001140740 Key Organics / BIONET

4-(chloromethyl)-5-[(2-chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole

  • MDL number MFCD00664820
  • Cas Number : 318289-54-2
  • Molecular Weight : 349.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 69 - 70
Iupac Name 4-(chloromethyl)-5-[(2-chlorophenyl)sulfanyl]-1-methyl-3-phenyl-1H-pyrazole
Inchi Code 1S/C17H14Cl2N2S/c1-21-17(22-15-10-6-5-9-14(15)19)13(11-18)16(20-21)12-7-3-2-4-8-12/h2-10H,11H2,1H3
Inchi Key FHGWSKBENMLOTK-UHFFFAOYSA-N

Description

Melting Point 69 - 70
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78925

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/5K-362S_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart