KEY001140873 Key Organics / BIONET

3,5-dioxo-2-[3-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

  • MDL number MFCD00274940
  • Cas Number : 121065-97-2
  • Molecular Weight : 282.182

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173 - 175
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79073

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5K-915_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart