KEY001140877 Key Organics / BIONET

2-(4-fluorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

  • MDL number MFCD00794272
  • Cas Number : 338982-40-4
  • Molecular Weight : 232.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242 - 244
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79077

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5K-921_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart