KEY001140914 Key Organics / BIONET

ethyl 4-(2-chloro-6-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD00664286
  • Cas Number : 299920-90-4
  • Molecular Weight : 311.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200 - 202
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79117

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5L-027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart