KEY001140935 Key Organics / BIONET

ethyl 4-(2,4-dichlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD00664292
  • Cas Number : 303136-96-1
  • Molecular Weight : 328.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 190 - 192
Iupac Name ethyl 4-(2,4-dichlorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate
Inchi Code 1S/C15H15Cl2NO3/c1-3-21-15(20)14-8(2)18-13(19)7-11(14)10-5-4-9(16)6-12(10)17/h4-6,11H,3,7H2,1-2H3,(H,18,19)
Inchi Key JLIPNJXXCMNOJM-UHFFFAOYSA-N

Description

Melting Point 190 - 192
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79138

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5L-055_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart