KEY001140936 Key Organics / BIONET

ethyl 4-(2-fluorophenyl)-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD00664293
  • Cas Number : 303139-43-7
  • Molecular Weight : 277.295

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150 - 152
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79139

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5L-056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart