KEY001141007 Key Organics / BIONET

6-[(4-chlorophenyl)sulfanyl]pyridine-3-carbaldehyde

  • MDL number MFCD00974900
  • Cas Number : 328547-40-6
  • Molecular Weight : 249.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 61.5 - 63.5
Iupac Name 6-[(4-chlorophenyl)sulfanyl]nicotinaldehyde
Inchi Code 1S/C12H8ClNOS/c13-10-2-4-11(5-3-10)16-12-6-1-9(8-15)7-14-12/h1-8H
Inchi Key GDUCSOHYPPLYFQ-UHFFFAOYSA-N

Description

Melting Point 61.5 - 63.5
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79216

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5L-356S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart