KEY001141441 Key Organics / BIONET

2-chloro-6-cyclopropyl-4-(trifluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD01316492
  • Cas Number : 478049-48-8
  • Molecular Weight : 246.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-6-cyclopropyl-4-(trifluoromethyl)nicotinonitrile
Inchi Code 1S/C10H6ClF3N2/c11-9-6(4-15)7(10(12,13)14)3-8(16-9)5-1-2-5/h3,5H,1-2H2
Inchi Key PTRGOELLIOIXBJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Liquid,Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79684

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/5N-333S_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart