KEY001141479 Key Organics / BIONET

6-tert-butyl-2-hydroxy-4-(trifluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD00843141
  • Cas Number : 478049-68-2
  • Molecular Weight : 244.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202 - 203
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79722

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-373S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart