KEY001141645 Key Organics / BIONET

thieno[2,3-d]pyrimidin-4-amine

  • MDL number MFCD01316189
  • Cas Number : 14080-56-9
  • Molecular Weight : 151.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234 - 236
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79893

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-841_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart