KEY001141658 Key Organics / BIONET

2-(4-chlorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile

  • MDL number MFCD01316193
  • Cas Number : 20932-04-1
  • Molecular Weight : 248.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 220 - 224
Iupac Name 2-(4-chlorophenyl)-3,5-dioxo-2,3,4,5-tetrahydro-1,2,4-triazine-6-carbonitrile
Inchi Code 1S/C10H5ClN4O2/c11-6-1-3-7(4-2-6)15-10(17)13-9(16)8(5-12)14-15/h1-4H,(H,13,16,17)
Inchi Key YPLNUPQUGLQZPI-UHFFFAOYSA-N

Description

Melting Point 220 - 224
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79906

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5N-919_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart