KEY001141750 Key Organics / BIONET

1-(4-chlorophenyl)-4-oxo-1,4-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD01871546
  • Cas Number : 147920-34-1
  • Molecular Weight : 250.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230 - 234
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80029

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5P-522S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart