KEY001141781 Key Organics / BIONET

methyl 1-(6-methylpyridin-2-yl)-1H-imidazole-4-carboxylate

  • MDL number MFCD02082748
  • Cas Number : 220965-34-4
  • Molecular Weight : 217.228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186 - 189
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80077

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5P-703_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart