KEY001141855 Key Organics / BIONET

6-tert-butyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one

  • MDL number MFCD00466456
  • Cas Number : 66698-66-6
  • Molecular Weight : 184.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176 - 178
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80189

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0257_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart