KEY001141932 Key Organics / BIONET

N-{3-[(2-chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetamide

  • MDL number MFCD02186592
  • Cas Number : 453557-52-3
  • Molecular Weight : 282.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 93
Iupac Name N-{3-[(2-chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetamide
Inchi Code 1S/C12H11ClN2O2S/c1-8(16)15-9-3-2-4-10(5-9)17-7-11-6-14-12(13)18-11/h2-6H,7H2,1H3,(H,15,16)
Inchi Key PKTGMAZTFQFXEI-UHFFFAOYSA-N

Description

Melting Point 93
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80296

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/5R-0679_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart