KEY001141934 Key Organics / BIONET

2-{4-[(2-chloro-1,3-thiazol-5-yl)methoxy]phenyl}acetonitrile

  • MDL number MFCD02186594
  • Cas Number : 672950-06-0
  • Molecular Weight : 264.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68 - 69
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80298

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0681_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart