KEY001143230 Key Organics / BIONET

2-(6-chloropyridazin-3-yl)-2-(pyridin-2-yl)acetonitrile

  • MDL number MFCD00140706
  • Cas Number : 338779-25-2
  • Molecular Weight : 230.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233 - 235
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81308

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6D-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart