KEY001143317 Key Organics / BIONET

1-{[3,5-bis(trifluoromethyl)phenyl]methyl}-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00243697
  • Cas Number : 338781-55-8
  • Molecular Weight : 365.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 169
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81401

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6E-311S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart