KEY001143372 Key Organics / BIONET

6-oxo-1-{[4-(trifluoromethyl)phenyl]methyl}-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00243721
  • Cas Number : 338783-75-8
  • Molecular Weight : 297.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 251 - 253
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81459

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6E-378S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart