KEY001143544 Key Organics / BIONET

3-amino-1H-indole-2-carbohydrazide

  • MDL number MFCD00141131
  • Cas Number : 110963-29-6
  • Molecular Weight : 190.206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 199
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81690

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6F-945_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart