KEY001143548 Key Organics / BIONET

ethyl 3-imino-3-{[(methoxycarbonyl)amino]amino}propanoate

  • MDL number MFCD00664782
  • Cas Number : 338791-53-0
  • Molecular Weight : 203.198

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 122
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81696

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6F-952_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart