KEY001143593 Key Organics / BIONET

(3E)-1,1,1-trifluoro-4-methoxypent-3-en-2-one

  • MDL number MFCD00173110
  • Cas Number : 102145-82-4
  • Molecular Weight : 168.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 41@9mm Hg
Iupac Name (3E)-1,1,1-trifluoro-4-methoxy-3-penten-2-one
Inchi Code 1S/C6H7F3O2/c1-4(11-2)3-5(10)6(7,8)9/h3H,1-2H3/b4-3+
Inchi Key XJNIYNVIHHABIV-ONEGZZNKSA-N

Description

Melting Point 41@9mm Hg
Purity 95%
Storage temp. Store under N2
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81749

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/6G-061_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart