KEY001143595 Key Organics / BIONET

2-(6-chloropyridin-2-yl)-2-phenylacetonitrile

  • MDL number MFCD00172233
  • Cas Number : 24783-42-4
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 75 - 77
Iupac Name (6-chloro-2-pyridinyl)(phenyl)acetonitrile
Inchi Code 1S/C13H9ClN2/c14-13-8-4-7-12(16-13)11(9-15)10-5-2-1-3-6-10/h1-8,11H
Inchi Key GETLPXGITMSNKU-UHFFFAOYSA-N

Description

Melting Point 75 - 77
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81750

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6G-065_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart