KEY001143638 Key Organics / BIONET

2-(dimethylamino)-6-(trifluoromethyl)pyrimidin-4-ol

  • MDL number MFCD00171894
  • Cas Number : 55545-80-7
  • Molecular Weight : 207.156

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176 - 177
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81798

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6G-380S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart