KEY001143791 Key Organics / BIONET

3-[2-(1H-imidazol-1-yl)phenyl]-1H-1,2,4-triazole

  • MDL number MFCD00244275
  • Cas Number : 338793-57-0
  • Molecular Weight : 211.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 188 - 190
Iupac Name 3-[2-(1H-imidazol-1-yl)phenyl]-1H-1,2,4-triazole
Inchi Code 1S/C11H9N5/c1-2-4-10(16-6-5-12-8-16)9(3-1)11-13-7-14-15-11/h1-8H,(H,13,14,15)
Inchi Key MEOQWPMLKZKWFI-UHFFFAOYSA-N

Description

Melting Point 188 - 190
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81969

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6H-068_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart