KEY001143802 Key Organics / BIONET

3-benzamidothiophene-2-carboxylic acid

  • MDL number MFCD01316568
  • Cas Number : 478080-03-4
  • Molecular Weight : 247.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 200 - 201
Iupac Name 3-(benzoylamino)-2-thiophenecarboxylic acid
Inchi Code 1S/C12H9NO3S/c14-11(8-4-2-1-3-5-8)13-9-6-7-17-10(9)12(15)16/h1-7H,(H,13,14)(H,15,16)
Inchi Key AEBWJNJRYSGZCZ-UHFFFAOYSA-N

Description

Melting Point 200 - 201
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81979

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6H-304S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart