KEY001143804 Key Organics / BIONET

methyl 3-acetamidothiophene-2-carboxylate

  • MDL number MFCD00100044
  • Cas Number : 22288-79-5
  • Molecular Weight : 199.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94 - 95
Purity 95
Storage temp. RT
Physical Form White to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81981

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/6H-306S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart