KEY001143952 Key Organics / BIONET

(2E)-3-amino-3-(4-methylpiperazin-1-yl)prop-2-enenitrile

  • MDL number MFCD00664794
  • Cas Number : 136062-62-9
  • Molecular Weight : 166.228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 123
Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82145

Safety Information

signalword Warning
pcodes P411,P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6H-949_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart