KEY001143964 Key Organics / BIONET

ethyl 6-bromo-4-chloroquinoline-3-carboxylate

  • MDL number MFCD00173367
  • Cas Number : 206257-39-8
  • Molecular Weight : 314.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 102
Purity 97
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82158

Safety Information

signalword Warning
pcodes P232,P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6J-001_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart