KEY001143966 Key Organics / BIONET

ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

  • MDL number MFCD00173369
  • Cas Number : 26893-12-9
  • Molecular Weight : 285.222

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 336 dec
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82160

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6J-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart