KEY001144015 Key Organics / BIONET

ethyl 8-cyano-4-hydroxyquinoline-3-carboxylate

  • MDL number MFCD00173411
  • Cas Number : 77156-79-7
  • Molecular Weight : 242.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235 - 238
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82210

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6J-055_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart