KEY001144124 Key Organics / BIONET

5-[(4-methoxyphenyl)methyl]-2-phenylpyrimidine-4,6-diamine

  • MDL number MFCD00231625
  • Cas Number : 163590-00-9
  • Molecular Weight : 306.369

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181 - 183
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82342

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart