KEY001144128 Key Organics / BIONET

2-[(2,4-dichlorophenyl)methyl]propanedinitrile

  • MDL number MFCD00231628
  • Cas Number : 338965-08-5
  • Molecular Weight : 225.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68 - 70
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82346

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart