KEY001144179 Key Organics / BIONET

2-[cyano(2,4-dichlorophenyl)methyl]-6-fluorobenzonitrile

  • MDL number MFCD00231988
  • Cas Number : 338965-45-0
  • Molecular Weight : 305.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102 - 104
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82403

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-080_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart