KEY001144185 Key Organics / BIONET

3-[(4-fluorophenyl)methoxy]-2-methyl-1,4-dihydropyridin-4-one

  • MDL number MFCD00215663
  • Cas Number : 338965-51-8
  • Molecular Weight : 233.242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6K-086_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart