KEY001145388 Key Organics / BIONET

2-amino-2-(4-methylphenyl)acetic acid

  • MDL number MFCD00665357
  • Cas Number : 13227-01-5
  • Molecular Weight : 165.192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256 - 257
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82658

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6L-332S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart