KEY001145410 Key Organics / BIONET

6-[(2-chlorophenyl)sulfanyl]pyridine-3-carbaldehyde

  • MDL number MFCD00974921
  • Cas Number : 338967-22-9
  • Molecular Weight : 249.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65 - 66.5
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82684

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6L-365S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart