KEY001145477 Key Organics / BIONET

5-[2-(methylsulfanyl)phenyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00974931
  • Cas Number : 338968-03-9
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230 - 231
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/82768

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6L-608S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart