KEY001145781 Key Organics / BIONET

4-(6-chloro-2-phenylpyrimidin-4-yl)morpholine

  • MDL number MFCD00754198
  • Cas Number : 343373-72-8
  • Molecular Weight : 275.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128 - 130
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83094

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6N-344S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart