KEY001146019 Key Organics / BIONET

4-(4-chloro-3-fluorophenoxy)benzoic acid

  • MDL number MFCD01871572
  • Cas Number : 551921-67-6
  • Molecular Weight : 266.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83355

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-077_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart