KEY001146029 Key Organics / BIONET

2-phenyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD00170683
  • Cas Number : 42607-21-6
  • Molecular Weight : 209.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 179
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83372

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-115_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart