KEY001146169 Key Organics / BIONET

1-[2-(4-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]ethan-1-one

  • MDL number MFCD01871615
  • Cas Number : 54001-07-9
  • Molecular Weight : 251.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125 - 127
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83530

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6P-742_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart