KEY001146323 Key Organics / BIONET

3-amino-3-(4-methylphenyl)propanoic acid

  • MDL number MFCD01863249
  • Cas Number : 68208-18-4
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 226 - 228
Iupac Name 3-(4-methylphenyl)-beta-alanine
Inchi Code 1S/C10H13NO2/c1-7-2-4-8(5-3-7)9(11)6-10(12)13/h2-5,9H,6,11H2,1H3,(H,12,13)
Inchi Key XPDAKEOBPKFUAH-UHFFFAOYSA-N

Description

Melting Point 226 - 228
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/83754

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6R-0630_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart