KEY001146499 Key Organics / BIONET

methyl 4-amino-2-(4-benzylpiperazin-1-yl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD00139424
  • Cas Number : 343375-51-9
  • Molecular Weight : 332.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 174
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84612

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7C-049_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart