KEY001146501 Key Organics / BIONET

4-[(3,5-dimethyl-1,2-oxazol-4-yl)methyl]phenol

  • MDL number MFCD00215380
  • Cas Number : 75999-06-3
  • Molecular Weight : 203.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106 - 108
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84614

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7C-052_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart