KEY001146532 Key Organics / BIONET

4-(4-chlorophenyl)-2-oxo-2,5-dihydrofuran-3-carbonitrile

  • MDL number MFCD00099688
  • Cas Number : 39561-83-6
  • Molecular Weight : 219.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 230 - 232
Iupac Name 4-(4-chlorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Inchi Code 1S/C11H6ClNO2/c12-8-3-1-7(2-4-8)10-6-15-11(14)9(10)5-13/h1-4H,6H2
Inchi Key IJCBFQOOIOPFQG-UHFFFAOYSA-N

Description

Melting Point 230 - 232
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84647

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7D-002_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart