KEY001146538 Key Organics / BIONET

3-(benzyloxy)thiophene-2-carbohydrazide

  • MDL number MFCD00215166
  • Cas Number : 343375-80-4
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84654

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7D-010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart