KEY001146557 Key Organics / BIONET

methyl 3-{[3-(trifluoromethyl)phenyl]methoxy}thiophene-2-carboxylate

  • MDL number MFCD00202083
  • Cas Number : 343375-98-4
  • Molecular Weight : 316.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60 - 61
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84676

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7D-048_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart