KEY001146690 Key Organics / BIONET

1-benzyl-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00101801
  • Cas Number : 4332-79-0
  • Molecular Weight : 229.235

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205 - 207
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84818

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7E-330S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart