KEY001146726 Key Organics / BIONET

5-chloro-1-[(2,6-dichlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD00140281
  • Cas Number : 339009-02-8
  • Molecular Weight : 332.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 198 - 200
Iupac Name 5-chloro-1-(2,6-dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridinecarboxylic acid
Inchi Code 1S/C13H8Cl3NO3/c14-7-4-8(13(19)20)12(18)17(5-7)6-9-10(15)2-1-3-11(9)16/h1-5H,6H2,(H,19,20)
Inchi Key VGKXLEJSDBNNBG-UHFFFAOYSA-N

Description

Melting Point 198 - 200
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84869

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7E-396S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart